Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979935.1 | XP_027835736.1 | 1 - 2 | 122 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |