Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004383.4 | XP_004004432.1 | 7 - 8 | 964 | 106 | |
XM_012125908.3 | XP_011981298.1 | 7 - 8 | 937 | 106 | ||
XM_012125926.3 | XP_011981316.1 | 6 - 7 | 788 | 106 | ||
XM_012125943.3 | XP_011981333.1 | 7 - 8 | 974 | 106 | ||
XM_027964561.1 | XP_027820362.1 | 6 - 7 | 671 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |