Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092042.2 | XP_014947528.2 | 12 - 13 | 1100 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |