Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005657.4 | XP_004005706.2 | 2 - 3 | 999 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |