Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976059.1 | XP_027831860.1 | 4 - 5 | 329 | 109 |
Gene Symbol: LOC114117268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976060.1 | XP_027831861.1 | 1 - 2 | 127 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |