Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962427.1 | XP_027818228.1 | 3 - 4 | 1681 | 111 | |
XM_027962428.1 | XP_027818229.1 | 3 - 4 | 1693 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |