Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC114114002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967455.1 | XP_027823256.1 | 1 - 2 | 197 | 93 |
Gene Symbol: LOC101113191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964281.1 | XP_027820082.1 | 1 - 2 | 197 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |