Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016514.3 | XP_004016563.1 | 3 - 4 | 353 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |