Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016514.3 | XP_004016563.1 | 6 - 7 | 624 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |