Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004023360.4 | XP_004023409.3 | 3 - 4 | 455 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |