Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002506.4 | XP_004002555.1 | 4 - 5 | 750 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |