Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012181389.3 | XP_012036779.1 | 4 - 5 | 559 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |