Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABARAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012641.4 | XP_004012690.1 | 1 - 2 | 932 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |