Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979145.1 | XP_027834946.1 | 5 - 6 | 631 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |