Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101106665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964014.1 | XP_027819815.1 | 3 - 4 | 954 | 95 |