Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961479.1 | XP_027817280.1 | 1 - 2 | 716 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |