Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACTIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969881.1 | XP_027825682.1 | 4 - 5 | 932 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |