Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099486.2 | XP_014954972.2 | 1 - 2 | 138 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |