Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012147113.3 | XP_012002503.2 | 1 - 2 | 180 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |