Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004232.4 | XP_004004281.1 | 4 - 5 | 582 | 95 | |
XM_012120695.3 | XP_011976085.1 | 4 - 5 | 572 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |