Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012104360.2 | XP_011959750.1 | 2 - 3 | 262 | 91 | |
XM_012104361.2 | XP_011959751.1 | 3 - 4 | 256 | 91 | ||
XM_027962048.1 | XP_027817849.1 | 3 - 4 | 234 | 91 | ||
XM_027962049.1 | XP_027817850.1 | 3 - 4 | 316 | 91 | ||
XM_027962050.1 | XP_027817851.1 | 4 - 5 | 293 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |