Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980157.1 | XP_027835958.1 | 8 - 9 | 1293 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |