Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4HTM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957663.1 | XP_027813464.1 | 8 - 9 | 1283 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |