Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099408.2 | XP_014954894.1 | 4 - 5 | 759 | 65 | |
XM_027976014.1 | XP_027831815.1 | 4 - 5 | 414 | 65 |