Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184578.3 | XP_012039968.1 | 1 | 3391 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |