Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102208.2 | XP_014957694.1 | 3 - 4 | 214 | 93 | |
XM_015102210.2 | XP_014957696.1 | 3 - 4 | 252 | 93 |