Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013794.4 | XP_004013843.2 | 4 - 5 | 2233 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |