Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LITAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104081.2 | XP_014959567.1 | 3 - 4 | 441 | 102 | |
XM_027961902.1 | XP_027817703.1 | 3 - 4 | 704 | 102 | ||
XM_027961903.1 | XP_027817704.1 | 3 - 4 | 1222 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |