Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978237.1 | XP_027834038.1 | 13 - 14 | 3467 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |