Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HGH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184243.2 | XP_012039633.2 | 5 - 6 | 1089 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |