Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006137.4 | XP_004006186.2 | 4 - 5 | 732 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |