Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008920.3 | XP_004008969.1 | 1 - 2 | 1763 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |