Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012781.4 | XP_004012830.2 | 1 - 2 | 576 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |