Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOATZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016008.4 | XP_004016057.2 | 5 - 6 | 887 | 155 | |
XM_015100907.2 | XP_014956393.2 | 5 - 6 | 715 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |