Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016999.3 | XP_004017048.2 | 3 - 4 | 349 | 127 | |
XM_012096943.2 | XP_011952333.2 | 3 - 4 | 351 | 127 | ||
XM_012096945.3 | XP_011952335.1 | 2 - 3 | 280 | 127 | ||
XM_027980074.1 | XP_027835875.1 | 4 - 5 | 494 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |