Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104813.2 | XP_014960299.2 | 1 - 2 | 183 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |