Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961923.1 | XP_027817724.1 | 3 - 4 | 495 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |