Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964876.1 | XP_027820677.1 | 1 - 2 | 1587 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |