Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101581.2 | XP_014957067.1 | 2 - 3 | 495 | 73 | |
XM_027956419.1 | XP_027812220.1 | 2 - 3 | 2846 | 73 | ||
XM_027956420.1 | XP_027812221.1 | 2 - 3 | 212 | 73 | ||
XM_027956421.1 | XP_027812222.1 | 3 - 4 | 234 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |