Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975901.1 | XP_027831702.1 | 4 - 5 | 1531 | 117 | |
XM_027975909.1 | XP_027831710.1 | 4 - 5 | 1192 | 117 | ||
XM_027975912.1 | XP_027831713.1 | 4 - 5 | 1216 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |