Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017798.4 | XP_004017847.1 | 4 - 5 | 497 | 106 | |
XM_012098586.3 | XP_011953976.1 | 4 - 5 | 581 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |