Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100399.2 | XP_014955885.2 | 1 | 542 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |