Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012180163.3 | XP_012035553.2 | 3 - 4 | 687 | 72 | |
XM_012180164.3 | XP_012035554.2 | 3 - 4 | 687 | 72 | ||
GenBank mRNA | Y09830.1 | - | 190 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |