Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012186587.3 | XP_012041977.1 | 10 - 11 | 1506 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |