Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019415.4 | XP_004019464.2 | 3 - 4 | 735 | 106 | |
XM_027960121.1 | XP_027815922.1 | 2 - 3 | 468 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |