Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012098549.3 | XP_011953939.1 | 2 - 3 | 176 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |