Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101110178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980021.1 | XP_027835822.1 | 4 - 5 | 486 | 89 | |
XM_027980022.1 | XP_027835823.1 | 4 - 5 | 486 | 89 | ||
XM_027980023.1 | XP_027835824.1 | 4 - 5 | 522 | 89 | ||
XM_027980024.1 | XP_027835825.1 | 4 - 5 | 486 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |