Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013801.4 | XP_004013850.2 | 3 - 4 | 789 | 91 | |
XM_012187497.3 | XP_012042887.2 | 3 - 4 | 607 | 91 | ||
XM_027975875.1 | XP_027831676.1 | 3 - 4 | 607 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |