Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099215.2 | XP_014954701.1 | 1 - 2 | 158 | 52 | |
XM_027975635.1 | XP_027831436.1 | 1 - 2 | 156 | 52 | ||
XM_027975636.1 | XP_027831437.1 | 1 - 2 | 155 | 52 |