Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961648.1 | XP_027817449.1 | 4 - 5 | 428 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |